Non-return valves
7,6 - 10 mm ID
Catalog # 229-3330
Supplier: KARTELL
Jump to:
Product Details & Documents
PP, translucent. Prevent pressure loss and backflow. Maximum working pressure 2 bar and minimum working pressure 0,07 bar at room temperature (20 °C).
- Excellent chemical resistance
- Excellent thermal resistance, autoclavable
- Tapered ends to ensure that tubes are held firmly
Documents
CoC Non-Return Valves.pdf
Conformity declaration
Specifications
- Velikost hadice/trubice:7,6 - 10 mm ID