Microplates, 96-well
10, White, LUMITRAC™ 200 medium binding, 40 - 340 µl, 96, -, 392 µl
Katalogové číslo 736-0228
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifikace
- Poč. jamek:96
- Doporučený pracovní objem:40 - 340 µl
- Sterilní:-
- Popis:White, LUMITRAC™ 200 medium binding
- Sekvence:10
- Celkový objem:392 µl
- Cat. No.:736-0228
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact