Orders LinkContactUsLinkComponent
Wire triangles
Wire triangles
02, With smooth earthenware pipes, tube length 60 mm
Katalogové číslo HECH41065006
Wire triangles
Katalogové číslo HECH41065006
Wire triangles
Wire triangles
02, With smooth earthenware pipes, tube length 60 mm
Přejít na:
Price
153,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These wire triangles with plain clay tubes are made of galvanised steel wire.
Specifikace
  • Popis:
    With smooth earthenware pipes, tube length 60 mm
  • Sekvence:
    02