Orders LinkContactUsLinkComponent
Wire triangles
Wire triangles
01, With smooth earthenware pipes, tube length 40 mm
Catalog # HECH41065004
Wire triangles
Catalog # HECH41065004
Wire triangles
Wire triangles
01, With smooth earthenware pipes, tube length 40 mm
Price
138,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These wire triangles with plain clay tubes are made of galvanised steel wire.
Specifications
  • Popis:
    With smooth earthenware pipes, tube length 40 mm
  • Sekvence:
    01