Orders LinkContactUsLinkComponent
Forceps
Forceps
Blunt, 253 mm
Catalog # 232-2003
Supplier:   KARTELL
Forceps
Catalog # 232-2003
Supplier:  KARTELL
Forceps
Forceps
Blunt, 253 mm
Price
2 030,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance, not suitable for use with strong acids and solvents
  • Autoclavable
  • Robust, pliable, self-sprung
Specifications
  • Type:
    Blunt
  • Délka:
    253 mm