Forceps
Blunt, 118 mm
Katalogové číslo 232-2000
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
- Good chemical resistance, not suitable for use with strong acids and solvents
- Autoclavable
- Robust, pliable, self-sprung
Specifikace
- Type:Blunt
- Délka:118 mm