Orders LinkContactUsLinkComponent
Quartz control plates
Quartz control plates
02, Standardní křemenná ovládací deska +15 ° Z, +5,2 °A, 1
Katalogové číslo 635-0023
Dodavatel:   BELLINGHAM STANLEY
Quartz control plates
Katalogové číslo 635-0023
Dodavatel:  BELLINGHAM STANLEY
Číslo CAS:  
Quartz control plates
Quartz control plates
02, Standardní křemenná ovládací deska +15 ° Z, +5,2 °A, 1
Přejít na:
Číslo CAS:  
Price
56 100,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
 : Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
 : Sadlles can be ordered in option.
 : Supplied with a certificate of verification in a plastic storage case.
Specifikace
Specifikace
  • Bal.:
    1
  • Popis:
    Standardní křemenná ovládací deska +15 ° Z, +5,2 °A
  • Sekvence:
    02
Specifikace