Pipeta Pasteur nesterilní graduace 2ml (500 ks) 1 * 500 KS
Catalog # 612-0009
Dodavatel: Alpha Laboratories
Pipeta Pasteur nesterilní graduace 2ml (500 ks) 1 * 500 KS
Catalog # 612-0009
Dodavatel: Alpha Laboratories
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Dodavatel: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Dodavatel: Corning
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
VWR®, Vzorkovací lžíce
Dodavatel: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Dodavatel: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Dodavatel: VWR Collection
Safety wash bottles, narrow neck, right-to-understand, Nalgene®
Dodavatel: Thermo Fisher Scientific
VWR®, Zkumavky pro PCR, 0,2 ml
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection