Measuring beaker with handle
, 2000 ml,
Katalogové číslo NICKJUGS2000H
Dodavatel: NICKEL ELECTRO
Přejít na:
Product Details & Documents
Stainless steel, rust-proof.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for fast, easy cleaning
Conical, without rim, with broad base and textured graduations.
Specifikace
- Kapacita:2000 ml