Orders LinkContactUsLinkComponent
Cork borers
Cork borers
Cork borers (12), 5 − 18 mm
Catalog # 217-0705
Supplier:   BOCHEM
Cork borers
Catalog # 217-0705
Supplier:  BOCHEM
Cork borers
Cork borers
Cork borers (12), 5 − 18 mm
Price
6 470,00 Kč
Log in for availability
1 * 1 SET
Product Details & Documents
Cork borers made of hard coated nickel-plated brass.
Delivery information: Include a cleaning needle.
Specifications
  • Ø:
    5 − 18 mm
  • Popis:
    Cork borers (12)