Cork borers
Cork borers (12), 5 − 18 mm
Catalog # 217-0705
Supplier: BOCHEM
Jump to:
Product Details & Documents
Cork borers made of hard coated nickel-plated brass.
Delivery information: Include a cleaning needle.
Specifications
- Ø:5 − 18 mm
- Popis:Cork borers (12)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.