Orders LinkContactUsLinkComponent
Cork borers
Cork borers
Cork borers (6), 5 − 11 mm
  217-0704
 :   BOCHEM
Cork borers
  217-0704
 :  BOCHEM
Cork borers
Cork borers
Cork borers (6), 5 − 11 mm
3 600,00 Kč
1 * 1 SET
Cork borers made of hard coated nickel-plated brass.
Delivery information: Include a cleaning needle.
  • Ø:
    5 − 11 mm
  • Popis:
    Cork borers (6)