Orders LinkContactUsLinkComponent
Slide cabinets
Slide cabinets
002, Slide cabinet for six drawers, 510×458×130 mm, 5496
Catalog # LPIT191084
Supplier:   LP ITALIANA
Slide cabinets
Catalog # LPIT191084
Supplier:  LP ITALIANA
Slide cabinets
Slide cabinets
002, Slide cabinet for six drawers, 510×458×130 mm, 5496
Price
16 530,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Plastic cabinets with drawers for microscopy and histology products.
Specifications
  • Počet sklíček:
    5496
  • Popis:
    Slide cabinet for six drawers
  • Sekvence:
    002
  • Š×H×V:
    510×458×130 mm