Disposable scalpels, Minor
08, No. 11
Supplier: SWANN MORTON
Jump to:
Product Details & Documents
These non sterile disposable scalpels consist of a stainless steel blade and a PS handle.
- 'Thumb' scalpels with a protective guard
- Ideal for surgical applications
Please note: The protective guard is NOT compatible with a standard metal handle and blade.
Ordering information: Available in packets of 10 or bulk boxes of 100.
Specifications
- Type:No. 11
- Sekvence:08
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.