Orders LinkContactUsLinkComponent
Disposable scalpels, Minor
Disposable scalpels, Minor
08, No. 11
Catalog # SWAN2373
Supplier:   SWANN MORTON
Disposable scalpels, Minor
Catalog # SWAN2373
Supplier:  SWANN MORTON
Disposable scalpels, Minor
Disposable scalpels, Minor
08, No. 11
Price
4 860,00 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
These non sterile disposable scalpels consist of a stainless steel blade and a PS handle.
  • 'Thumb' scalpels with a protective guard
  • Ideal for surgical applications
Please note: The protective guard is NOT compatible with a standard metal handle and blade.
Ordering information: Available in packets of 10 or bulk boxes of 100.
Specifications
  • Type:
    No. 11
  • Sekvence:
    08