Orders LinkContactUsLinkComponent
Tissue forceps, Adson / Micro-Adson
Tissue forceps, Adson / Micro-Adson
04, Micro-Adson, delicate, Straight, 120 mm
Katalogové číslo HAMML-HSC232-12
Dodavatel:   HAMMACHER KARL
Tissue forceps, Adson / Micro-Adson
Katalogové číslo HAMML-HSC232-12
Dodavatel:  HAMMACHER KARL
Tissue forceps, Adson / Micro-Adson
Tissue forceps, Adson / Micro-Adson
04, Micro-Adson, delicate, Straight, 120 mm
Přejít na:
Price
1 630,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Delicate forceps made from stainless steel.
Specifikace
  • Type:
    Straight
  • Popis:
    Micro-Adson, delicate
  • Délka:
    120 mm
  • Sekvence:
    04