Orders LinkContactUsLinkComponent
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Three-way stopcocks, Červený
Katalogové číslo 720-2574
Dodavatel:   B. Braun
Multi-way stopcocks, Discofix®
Katalogové číslo 720-2574
Dodavatel:  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Three-way stopcocks, Červený
Přejít na:
Price
2 020,00 Kč
Přihlásit se pro dostupnost
1 * 50 KS
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Specifikace
  • Barva:
    Červený
  • Popis:
    Three-way stopcocks