Orders LinkContactUsLinkComponent
Spore suspensions
Spore suspensions
03, Bacillus atrophaeus, cell line 9372: spore suspension (24 month shelf life), population 108/0,1 ml - steam sterilisation process
Katalogové číslo NAMSVBA-108
Dodavatel:   CROSSTEX
Spore suspensions
Katalogové číslo NAMSVBA-108
Dodavatel:  CROSSTEX
Spore suspensions
03, Bacillus atrophaeus, cell line 9372: spore suspension (24 month shelf life), population 108/0,1 ml - steam sterilisation process
Přejít na:
Price
70 530,00 Kč
Přihlásit se pro dostupnost
1 * 10 mL
Product Details & Documents
Documents
Specifikace
  • Popis:
    Bacillus atrophaeus, cell line 9372: spore suspension (24 month shelf life), population 108/0,1 ml - steam sterilisation process
  • Sekvence:
    03