Orders LinkContactUsLinkComponent
Stoppers, press down, VacuCap®
Stoppers, press down, VacuCap®
Zelená, 16 mm
Katalogové číslo 217-0104
Dodavatel:   Simport Scientific
Stoppers, press down, VacuCap®
Katalogové číslo 217-0104
Dodavatel:  Simport Scientific
Stoppers, press down, VacuCap®
Stoppers, press down, VacuCap®
Zelená, 16 mm
Přejít na:
Price
684,00 Kč
Přihlásit se pro dostupnost
1 * 1.000 KS
Product Details & Documents
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
  • Disposable seals are an inexpensive alternative to standard seals
  • Double flanged seal for an impermeable, secure fit
  • Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
Specifikace
  • Ø dno:
    16 mm
  • Barva:
    Zelená