Orders LinkContactUsLinkComponent
Staining racks
Staining racks
03, Rack, without dividers, 435×85 mm
Catalog # DELTS-003
Supplier:   DELTALAB
Staining racks
Catalog # DELTS-003
Supplier:  DELTALAB
Staining racks
03, Rack, without dividers, 435×85 mm
Price
1 320,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel staining racks for microscope slides with and without dividers.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Popis:
    Rack, without dividers, 435×85 mm
  • Sekvence:
    03