Orders LinkContactUsLinkComponent
Measuring spoons
Measuring spoons
01, Measuring spoon, molded plastic cup, black, 0,05 g
Catalog # HACH49200
Supplier:   Hach
Measuring spoons
Catalog # HACH49200
Supplier:  Hach
Measuring spoons
Measuring spoons
01, Measuring spoon, molded plastic cup, black, 0,05 g
Price
168,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Molded plastic cup. Weight based on sodium chloride.
Specifications
  • Kapacita:
    0,05 g
  • Popis:
    Measuring spoon, molded plastic cup, black
  • Sekvence:
    01