Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
5 ml, 0,05 ml, 0,015 ± ml
Catalog # BURK9695-1005
Supplier:   BURKLE
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Catalog # BURK9695-1005
Supplier:  BURKLE
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
5 ml, 0,05 ml, 0,015 ± ml
Price
1 940,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3, class B.
  • Adjusted for Ex
  • Blue ring graduations, Schellbach stripe
  • Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet
Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Certifications: Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Specifications
  • Kapacita:
    5 ml
  • Dělení:
    0,05 ml
  • Tolerance:
    0,015 ± ml
Accessories
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • BURK9695-0040
    Spare nozzle, PP
    Log in for availability
    1 * 1 KS - 79,10 Kč
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • BURK9695-0528
    Cap without feed tube for 1000 ml bottle
    Log in for availability
    1 * 1 KS - 47,70 Kč