Orders LinkContactUsLinkComponent
Micro pipettes, disposable, Microcaps®
32, Bulb dispenser
Catalog # 612-4865
Supplier:   DRUMMOND
Micro pipettes, disposable, Microcaps®
Catalog # 612-4865
Supplier:  DRUMMOND
Micro pipettes, disposable, Microcaps®
Micro pipettes, disposable, Microcaps®
32, Bulb dispenser
Price
165,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.
For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.
 : Delivery includes 100 pipettes and one bulb dispenser.
Specifications
  • Type:
    Bulb dispenser
  • Sekvence:
    32
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • DRUM1-000-9003
    01
    Silicone tips for Microcaps®
    Log in for availability
    1 * 3 KS - 168,00 Kč