Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
13, 2000 ml, 29/32 NS
Katalogové číslo BIBB443065N
Dodavatel:   SciLabware
Separating funnels, Gilson
Katalogové číslo BIBB443065N
Dodavatel:  SciLabware
Separating funnels, Gilson
Separating funnels, Gilson
13, 2000 ml, 29/32 NS
Přejít na:
Price
7 130,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Specifikace
  • Rozměry zábrusu:
    29/32 NS
  • Kapacita:
    2000 ml
  • Sekvence:
    13