Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
09, 125 ml, 19/26 NS
Katalogové číslo BIBB443042N
Dodavatel:   SciLabware
Separating funnels, Gilson
Katalogové číslo BIBB443042N
Dodavatel:  SciLabware
Separating funnels, Gilson
Separating funnels, Gilson
09, 125 ml, 19/26 NS
Přejít na:
Price
2 700,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Specifikace
  • Rozměry zábrusu:
    19/26 NS
  • Kapacita:
    125 ml
  • Sekvence:
    09