Orders LinkContactUsLinkComponent
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints
100 ml, 130 mm, 24/29 NS
Katalogové číslo BIBBFF100/3M
Dodavatel:   SciLabware
Flasks, flat bottom with Quickfit® ground joints
Katalogové číslo BIBBFF100/3M
Dodavatel:  SciLabware
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints
100 ml, 130 mm, 24/29 NS
Přejít na:
Price
4 880,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
PYREX®, borosilicate glass, clear, with medium necks.
  • Very good chemical resistance
  • High temperature resistance
  • With longer neck than standard of flasks
Flat base allows use with hotplates. With labelling area.
Specifikace
  • Velikost objímky:
    24/29 NS
  • Kapacita:
    100 ml
  • Výška:
    130 mm