Orders LinkContactUsLinkComponent
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
, , , S 13/5, -
Catalog # 201-8203
Supplier:   SciLabware
Spherical ground joints, Quickfit®
Catalog # 201-8203
Supplier:  SciLabware
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
, , , S 13/5, -
Price
5 290,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Specifications
  • Kulička:
    S 13/5
  • Zásuvka:
    -