Orders LinkContactUsLinkComponent
Slide box
Slide box
Průhledná/bílá, PS, 100, 38×360×100 mm
Catalog # 631-9750
Supplier:   KARTELL
Slide box
Catalog # 631-9750
Supplier:  KARTELL
Slide box
Slide box
Průhledná/bílá, PS, 100, 38×360×100 mm
Price
1 350,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Each cavity is numbered for storage in drawers.
W×D×H: 38×360×100 mm
Specifications
  • Počet sklíček:
    100
  • Barva:
    Průhledná/bílá
  • Materiál:
    PS
  • Š×H×V:
    38×360×100 mm