Wire baskets
08, 300×200 mm
Katalogové číslo 216-2846
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Stainless steel.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Specifikace
- Ø×H:300×200 mm
- Sekvence:08