Screw caps, bakelite
22, with TRISEAL seal, 28 mm, Černá
215-2698
: GRAVIS
- Závit:28 mm
- Barva:Černá
- Popis:with TRISEAL seal
- Sekvence:22
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
#: APOSBIA1493-1G
: Apollo Scientific
Vápník 1000 mg/L v 0,5 M kyselině dusičné (from Ca(NO₃)₂) Certipur® standard pro AAS, Supelco®
#: 1.19778.0100
: Merck
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (by titrimetric analysis), white powder
#: 42721.18
: Thermo Fisher Scientific