Orders LinkContactUsLinkComponent
Snap plug caps, WHEATON®
Snap plug caps, WHEATON®
04, PE green, 8 mm
Catalog # WHEA242806
Supplier:   DWK Life Sciences
Snap plug caps, WHEATON®
Catalog # WHEA242806
Supplier:  DWK Life Sciences
Snap plug caps, WHEATON®
Snap plug caps, WHEATON®
04, PE green, 8 mm
Price
2 690,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
Specifications
  • Ø:
    8 mm
  • Type:
    PE green
  • Sekvence:
    04