Orders LinkContactUsLinkComponent
Screw caps, bakelite
Screw caps, bakelite
17, with rubber liner, 18 mm, Černá
Catalog # 215-2696
Supplier:   GRAVIS
Screw caps, bakelite
Catalog # 215-2696
Supplier:  GRAVIS
Screw caps, bakelite
Screw caps, bakelite
17, with rubber liner, 18 mm, Černá
Price
1 210,00 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
Documents
Specifications
  • Závit:
    18 mm
  • Barva:
    Černá
  • Popis:
    with rubber liner
  • Sekvence:
    17