Bottle Carriers
Bottle carrier, HDPE, for food with closed floor, Šedá, Up to eight bottles, 390×110 mm, 95 mm
Katalogové číslo BURK0361-0008
Dodavatel: BURKLE
Přejít na:
Product Details & Documents
Made of PP or HDPE.
- Bottle transport container (PP) for up to 6 bottles, bottle transport box for up to 8 bottles
- Bottle transport box (HDPE): suitable for food; With impermeable base suitable as a collecting basin
Documents
BURK0361-0006_TDS_EN
Technický list
Specifikace
- Určeno pro:Up to eight bottles
- Barva:Šedá
- Popis:Bottle carrier, HDPE, for food with closed floor
- Velikost přihrádky:390×110 mm
- Max. Ø nádoby:95 mm