Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
06, Vacuum stopcocks, one way, straight, 2,5 mm, 14,5 NS
Katalogové číslo LENZ02901402
Dodavatel:   Lenz Laborglas
Vacuum stopcocks
Katalogové číslo LENZ02901402
Dodavatel:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
06, Vacuum stopcocks, one way, straight, 2,5 mm, 14,5 NS
Přejít na:
Price
1 510,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifikace
  • Ø otvoru:
    2,5 mm
  • Popis:
    Vacuum stopcocks, one way, straight
  • Sekvence:
    06
  • Kohout:
    14,5 NS