Orders LinkContactUsLinkComponent
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
04, Head for 1 litre bottle, 60/37
Katalogové číslo LENZ05660207
Dodavatel:   Lenz Laborglas
Pycnometer heads with/without bottle
Katalogové číslo LENZ05660207
Dodavatel:  Lenz Laborglas
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
04, Head for 1 litre bottle, 60/37
Price
2 060,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Set, made of DURAN® borosilicate glass, consists of wide neck bottle with conical shoulder and NS joint, and pycnometer head.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for density determinations of bitumen-like and mineral materials.
Documents
Specifikace
  • NZ:
    60/37
  • Popis:
    Head for 1 litre bottle
  • Sekvence:
    04
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Bottle for pycnometer NS60/46, 1000 ml
    Bottle for pycnometer NS60/46, 1000 ml
    LENZ07690070
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 870,00 Kč