Process pH combination electrodes
25, pH combination electrode, with dome membrane, H 8481 HD, 1 - 14, 0...+100 °C, Sklo, Referid®, KPG annular gap, 12×225 mm, Screw plug head
Katalogové číslo SCOI285111072
Dodavatel: SI Analytics
Jump to:
Product Details & Documents
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Specifications
- Připojení:Screw plug head
- Ø×D:12×225 mm
- Type:H 8481 HD
- Popis:pH combination electrode, with dome membrane
- Materiál:Sklo
- Sekvence:25
- Diafragma:KPG annular gap
- Elektrolyt:Referid®
- Tepl.:0...+100 °C
- pH:1 - 14
- Cat. No.:SCOI285111072