Orders LinkContactUsLinkComponent
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with glass tip, 2 mm
Catalog # COWI016.9203-B
Supplier:   COWIE
Stopcocks, straight through, with tip
Catalog # COWI016.9203-B
Supplier:  COWIE
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with glass tip, 2 mm
Price
615,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Ø otvoru:
    2 mm
  • Popis:
    with glass tip