Volumetric overflow flasks, without stopper
06, 365 ml, Overflow flask
Katalogové číslo NEUB0372-10-365
Dodavatel: NEUBERT VOLUME GLASSWAERE
Volumetric overflow flasks, without stopper
Katalogové číslo NEUB0372-10-365
Dodavatel: NEUBERT VOLUME GLASSWAERE
Přejít na:
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
- Calibrated to deliver (TD, Ex)
- Cut and ground exactly at the volume-mark
- Without stopper
Specially designed for water analysis.
Specifikace
- Kapacita:365 ml
- Popis:Overflow flask
- Sekvence:06