Orders LinkContactUsLinkComponent
Sample spoons
Sample spoons
6, Sample spoon, 400 mm
Katalogové číslo 300-0021
Dodavatel:   BURKLE
Sample spoons
Katalogové číslo 300-0021
Dodavatel:  BURKLE
Sample spoons
Sample spoons
6, Sample spoon, 400 mm
Přejít na:
Price
483,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)
Specifikace
  • Popis:
    Sample spoon
  • Délka:
    400 mm
  • Sekvence:
    6