Orders LinkContactUsLinkComponent
Sample spoons
Sample spoons
3, Sample spoon, 210 mm
Katalogové číslo 300-0020
Dodavatel:   BURKLE
Sample spoons
Katalogové číslo 300-0020
Dodavatel:  BURKLE
Sample spoons
Sample spoons
3, Sample spoon, 210 mm
Přejít na:
Price
227,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)
Specifikace
  • Popis:
    Sample spoon
  • Délka:
    210 mm
  • Sekvence:
    3