Orders LinkContactUsLinkComponent
Azotometer/saccharometer
Azotometer/saccharometer
02, Azotometer/saccharometer, measuring tube alone, 10 ml, —, 1/10
Katalogové číslo NEUB0345-50-10
Azotometer/saccharometer
Katalogové číslo NEUB0345-50-10
Azotometer/saccharometer
02, Azotometer/saccharometer, measuring tube alone, 10 ml, —, 1/10
Price
1 580,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Azotometer/saccharometer made of borosilicate glass, with glass base.
Specifications
  • Popis:
    Azotometer/saccharometer, measuring tube alone, 10 ml
  • Sekvence:
    02
  • Dělení:
    1/10
  • Rozsah: