Orders LinkContactUsLinkComponent
Labels for slides
Labels for slides
006, Slide labels, Papír, Černá na bílé, 22×22 mm, On roll
Catalog # 631-9499
Labels for slides
Catalog # 631-9499
Labels for slides
Labels for slides
006, Slide labels, Papír, Černá na bílé, 22×22 mm, On roll
Price
875,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
These thin labels are suitable for use with microscopy slides.
  • Permanent adhesive
  • No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Specifications
  • Barva:
    Černá na bílé
  • Popis:
    Slide labels
  • Rozměry:
    22×22 mm
  • Materiál:
    Papír
  • Sekvence:
    006
  • Balené:
    On roll