Linen testers
5×, 31,6 mm, 30×30 mm
Catalog # 630-8327
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Satin matt chrome plated brass frame with silicate glass lens.
Specifications
- Ø:31,6 mm
- Zvětšení:5×
- Š×H:30×30 mm