Orders LinkContactUsLinkComponent
Folding precision magnifiers
Folding precision magnifiers
004, Folding magnifier, achromatic, 20×, 17 mm
Catalog # 630-8222
Supplier:   ESCHENBACH OPTIK
Folding precision magnifiers
Catalog # 630-8222
Supplier:  ESCHENBACH OPTIK
Folding precision magnifiers
Folding precision magnifiers
004, Folding magnifier, achromatic, 20×, 17 mm
Price
3 960,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
  • Protective case can serve as a handle
  • Provide distortion-free images
  • Chrome plated metal
Specifications
  • Ø:
    17 mm
  • Zvětšení:
    20×
  • Popis:
    Folding magnifier, achromatic
  • Sekvence:
    004