Orders LinkContactUsLinkComponent
Binocular magnifiers
Binocular magnifiers
001, Binocular magnifying lens with 250 mm working distance*, 2×
Katalogové číslo 630-8214
Dodavatel:   ESCHENBACH OPTIK
Binocular magnifiers
Katalogové číslo 630-8214
Dodavatel:  ESCHENBACH OPTIK
Binocular magnifiers
Binocular magnifiers
001, Binocular magnifying lens with 250 mm working distance*, 2×
Přejít na:
Price
4 700,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
 : Supplied with a protective box.
 : Not suitable for spectacle wearers.
Specifikace
  • Zvětšení:
  • Popis:
    Binocular magnifying lens with 250 mm working distance*
  • Sekvence:
    001