Connectors, PP
18, Coupling, male luer lock/female luer lock, —
Katalogové číslo 229-0521
Dodavatel: PIEPER FILTER
Přejít na:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Specifikace
- Hadice int.Ø:—
- Popis:Coupling, male luer lock/female luer lock
- Sekvence:18