Orders LinkContactUsLinkComponent
Connectors, PP
Connectors, PP
03, Connector, straight, male luer lock/tubing nipple, 3,2 mm
Catalog # 229-0517
Supplier:   PIEPER FILTER
Connectors, PP
Catalog # 229-0517
Supplier:  PIEPER FILTER
Connectors, PP
Connectors, PP
03, Connector, straight, male luer lock/tubing nipple, 3,2 mm
Price
643,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Hadice int.Ø:
    3,2 mm
  • Popis:
    Connector, straight, male luer lock/tubing nipple
  • Sekvence:
    03