Orders LinkContactUsLinkComponent
Wire baskets
Wire baskets
240×250×50 mm
Catalog # 216-7450
Supplier:   HUPFER METALLWERKE
Wire baskets
Catalog # 216-7450
Supplier:  HUPFER METALLWERKE
Wire baskets
Wire baskets
240×250×50 mm
Price
5 590,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Two flip handles
  • Base frame is stackable
  • Excellent thermal and chemical resistance
Wire steriliser strainer basket.
Documents
Specifications
  • Š×H×V:
    240×250×50 mm