Orders LinkContactUsLinkComponent
Holder for wipe boxes
Holder for wipe boxes
Large holder for one wipe box, 320×127×97 mm
Katalogové číslo 115-2106
Dodavatel:   HEATHROW
Holder for wipe boxes
Katalogové číslo 115-2106
Dodavatel:  HEATHROW
Holder for wipe boxes
Holder for wipe boxes
Large holder for one wipe box, 320×127×97 mm
Přejít na:
Price
1 270,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These clear acrylic holders fit boxes of small or large wipes from most major brands, for example KIMWIPES*.
  • Can be wall-mounted or put on a shelf or table
  • Smaller version attaches with double-back tape, larger version needs to be mounted with screws
  • Mounting materials included
Specifikace
  • Popis:
    Large holder for one wipe box
  • Š×H×V:
    320×127×97 mm