Orders LinkContactUsLinkComponent
Holder for wipe boxes
Holder for wipe boxes
Small holder for one wipe box, 118×122×84 mm
Catalog # 115-2105
Supplier:   HEATHROW
Holder for wipe boxes
Catalog # 115-2105
Supplier:  HEATHROW
Holder for wipe boxes
Holder for wipe boxes
Small holder for one wipe box, 118×122×84 mm
Price
564,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These clear acrylic holders fit boxes of small or large wipes from most major brands, for example KIMWIPES*.
  • Can be wall-mounted or put on a shelf or table
  • Smaller version attaches with double-back tape, larger version needs to be mounted with screws
  • Mounting materials included
Specifications
  • Popis:
    Small holder for one wipe box
  • Š×H×V:
    118×122×84 mm