Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
24, 3000 ml, 24/40 NS, coated
Katalogové číslo BUCH025263
Dodavatel:   BUCHI
Evaporating flasks
Katalogové číslo BUCH025263
Dodavatel:  BUCHI
Evaporating flasks
Evaporating flasks
24, 3000 ml, 24/40 NS, coated
Přejít na:
Price
7 020,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifikace
  • Velikost objímky:
    24/40 NS
  • Type:
    coated
  • Kapacita:
    3000 ml
  • Sekvence:
    24