Orders LinkContactUsLinkComponent
Spider evaporators
Spider with evaporating flasks, SJ24/40, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint
Catalog # BUCH011574
Supplier:   BUCHI
Spider evaporators
Catalog # BUCH011574
Supplier:  BUCHI
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ24/40, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint
Price
31 030,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifications
  • Popis:
    Spider with evaporating flasks, SJ24/40, glass, 50 ml
  • Typ:
    For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint